missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CSK Antibody (7C6), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59500MFV500
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CSK Monoclonal antibody specifically detects CSK in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Tekniske data
| CSK | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 7C6 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| C-Src kinase, c-src tyrosine kinase, EC 2.7.10, EC 2.7.10.2, MGC117393, Protein-tyrosine kinase CYL, tyrosine-protein kinase CSK | |
| Recombinant human CSK (1-450aa) purified from E. coli | |
| 0.1 mL | |
| Cell Cycle and Replication, Signal Transduction | |
| 1445 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion