Learn More
Epredia™ Richard-Allan Scientific™ Reusable Steel Knives, 16 cm, C Profile Steel
Enhance operation of Epredia™ CryoStar™ NX50 and NX70 Cryostats with Epredia™ CryoStar™ NX50 and NX70 Accessories.
Brand: Epredia™ 152020
2393.92 DKK valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Additional Details : Weight : 0.99000kg
Description
Accessories Include:
Specifications
Steel Knife | |
C | |
Epredia™ | |
16 cm |
Knives | |
Steel | |
No |