Learn More
Thermo Scientific™ Cryo Marker Pen Set
Product Code. 15289134 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Black
Multi-color
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15289134 Multi-color Pack of 4
15611105 Black Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15289134 Supplier Thermo Scientific™ Supplier No. 4000221
  • ONLINE EXCLUSIVE PRICE
    520.00 DKK
    466.31 DKK / Pakke med 4
    Spare 53.69 DKK
    10% Slukket
 Restricted Item
På lager
Label cryogenic vials to -200°C (-328°F) with the Thermo Scientific™ Cryo Marker Pen Set. The ink is fast-drying, nonsmearing and permanent.
Label cryogenic vials to -200°C (-328°F) with the Thermo Scientific™ Cryo Marker Pen Set. The ink is fast-drying, nonsmearing and permanent.
  • Multicolor set includes four fine-tip pens in black, blue, green and red
TRUSTED_SUSTAINABILITY
Specifications
Type Cryo Marker Pen Set
Temperature Range -200°C, -328°F
Color Multi-color
For Use With (Equipment) cryovials and boxes
Holds Vials