Få mere at vide
Tocris Bioscience™ CRT 0105950
Beskrivelse
CRT 0105950 is a potent LIMK1/2 inhibitor (IC50 values are 0.3 and 1 nM at LIMK 1 and 2, respectively). Disrupts microtubule organization, inhibits cofilin phosphorylation and increases α-tubulin acetylation in vitro.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-[[5-[3-(2-Chloro-4-methylphenyl)-4-pyridinyl]-2-thiazolyl]amino]phenol |
| CAS | 1661845-86-8 |
| Mængde | 10 mg |
| Molekylær formel | C21H16ClN3OS |
| Renhed | 98% |