Få mere at vide
CRT 0066854 hydrochloride
GREENER_CHOICE
PKCι and PKCζ inhibitor
Brand:  Tocris Bioscience 5922/10
Encompass
Produktkode 15923241
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
CRT 0066854 hydrochloride is a protein kinase C (PKC) inhibitor. Inhibits PKCι and PKCζ (IC50 values are 132 and 639 nM, respectively). Restores polarized morphogenesis in oncogene expressing MDCK cells, decreases colony formation in HeLa cells and inhibits lethal giant larvae 2 (LLGL2) phosphorylation.
Tekniske data
Tekniske data
(2 S)-3-Phenyl-N1-[5,6,7,8-tetrahydro-2-(4-pyridinyl)[1 ]benzothieno[2,3-d]pyrimidin-4-yl]-1,2-propanediamine dihydrochloride
2250019-91-9
>98%
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner