missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-Crotamiton, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Crotamiton was used to study the photocatalytic decomposition of crotamiton in aqueous solution using TiO2. N-Ethyl-o-crotonotoluidide (Crotamiton) was used to treat scabies and various pruritic dermatoses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Crotamiton was used to study the photocatalytic decomposition of crotamiton in aqueous solution using TiO2. N-Ethyl-o-crotonotoluidide (Crotamiton) was used to treat scabies and various pruritic dermatoses.
Solubility
Soluble in water (1:500), alcohol, methanol, ether, and ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Crotamiton was used to study the photocatalytic decomposition of crotamiton in aqueous solution using TiO2. N-Ethyl-o-crotonotoluidide (Crotamiton) was used to treat scabies and various pruritic dermatoses.
Solubility
Soluble in water (1:500), alcohol, methanol, ether, and ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Density | 0.987 |
Boiling Point | 153°C to 155°C (13 mmHg) |
Flash Point | 113°C (235°F) |
Linear Formula | CH3CH=CHCON(C2H5)C6H4CH3 |
Refractive Index | 1.5405 |
Quantity | 1 g |
Solubility Information | Soluble in water (1:500),alcohol,methanol,ether,and ethanol. |
Formula Weight | 203.28 |
Percent Purity | 97% |
Chemical Name or Material | Crotamiton |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'