missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Crosstide

Product Code. 16402779
Encompass_Preferred
Change view
Click to view available options
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
16402779 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402779 Supplier MedChemExpress Supplier No. HYP03151mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Crosstide is a peptide analog of glycogen synthase kinase α/β fusion protein sequence which is a substrate for Akt.

Specifications

Renhed 95.7%
CAS 171783-05-4
Indhold og opbevaring Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Molekylvægt (g/mol) 1164.23 g/mol
Produkttype Peptides
Mængde 1 mg
Forsendelsestilstand Room temperature
Opløselighed H2O : ≥ 50 mg/mL (42.95 mM)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.