Få mere at vide
Invitrogen™ CRNKL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570817
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 100%.
CRNKL1 is a protein coding gene.
Tekniske data
| CRNKL1 | |
| Polyclonal | |
| Unconjugated | |
| CRNKL1 | |
| 1200013P10Rik; 5730590A01Rik; C80326; CGI-201; CLF; Clf1; Crn; Crn, crooked neck-like 1; Crn, crooked neck-like 1 (Drosophila); Crnkl1; Crooked neck homolog; crooked neck pre-mRNA splicing factor 1; crooked neck pre-mRNA splicing factor-like 1; Crooked neck protein; crooked neck-like 1; crooked neck-like 1 protein; crooked neck-like protein 1; crooked neck-like protein 1-like; crooked neck-like protein 1-like protein; hCrn; MSTP021; SYF3; SYF3 pre-mRNA-splicing factor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51340 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BZJ0 | |
| CRNKL1 | |
| synthetic peptide directed towards the C-terminal region of human CRNKL1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |