missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CREB5 Antibody (PCRP-CREB5-1G8), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14073C
Additional Details : Weight : 0.00970kg
Description
CREB5 Monoclonal antibody specifically detects CREB5 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
CREB5 | |
Monoclonal | |
DyLight 650 | |
cAMP response element binding protein CRE-Bpa, cAMP response element-binding protein CRE-BPa, cAMP responsive element binding protein 5, cAMP responsive element binding protein 5 isoform alpha, cAMP-responsive element-binding protein 5, CREB-5, CREBPA, CRE-BPA, cyclic AMP-responsive element-binding protein 5, H_GS165L15.1 | |
Recombinant full-length human CREB5 protein (Uniprot: Q02930) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
PCRP-CREB5-1G8 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
9586 | |
Store at 4°C in the dark. | |
IgG2b κ |