Få mere at vide
Creatinine Zinc Chloride, TRC
Artikelnummer. 30137659
Skift visning
Mængde:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137659 10 g
30134358 50 g
30010157 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137659 Leverandør Toronto Research Chemicals Leverandørnr. TRCC78151310G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 62708-52-5
Molekylær formel 2 C4 H7 N3 O . Zn . 2 Cl
Molekylvægt (g/mol) 362.55
Synonym 4H-Imidazol-4-one, 2-amino-1,5-dihydro-1-methyl-, compd. with zinc chloride (ZnCl2) (2:1) (9CI, ACI), 4H-Imidazol-4-one, 2-amino-1,5-dihydro-1-methyl-, compd. with zinc chloride (2:1) (ZCI), Zinc chloride (ZnCl2), compd. with 2-amino-1,5-dihydro-1-methyl-4H-imidazol-4-one (1:2) (9CI), Zinc chloride, compd. with 2-amino-1,5-dihydro-1-methyl-4H-imidazol-4-one (1:2) (ZCI), Creatinine Zinc Chloride
IUPAC navn zinc;2-amino-3-methyl-4H-imidazol-5-one;dichloride
SMIL [Cl-].[Cl-].[Zn+2].CN1CC(=O)N=C1N.CN2CC(=O)N=C2N
Tekniske data
Kemisk navn eller materiale Creatinine zinc chloride
Mængde 10 g
InChI formel InChI=1S/2C4H7N3O.2ClH.Zn/c2*1-7-2-3(8)6-4(7)5;;;/h2*2H2,1H3,(H2,5,6,8);2*1H;/q;;;;+2/p-2
Formel vægt 359.9847