Få mere at vide
Creatinine-d3, TRC
Artikelnummer. 30362670
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30362670 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30362670 Leverandør Toronto Research Chemicals Leverandørnr. TRCC7815021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Creatinine-D3
CAS 143827-20-7
Renhedsgrad noter HPLC
Molekylær formel C4 2H3 H4 N3 O
Mængde 1 mg
Synonym 2-Amino-1,5-dihydro-1-(methyl-d3)-4H-imidazol-4-one, 1-(Methyl-d3)glycocyamidine, 1-(Methyl-d3)hydantoin-2-imide, 2-Amino-1-(methyl-d3)-1,5-dihydroimidazol-4-one, 2-Amino-1-(methyl-d3)imidazolin-4-one, NSC 13123-d3, TEGO Cosmo C 250-d3
InChI formel InChI=1S/C4H7N3O/c1-7-2-3(8)6-4(7)5/h2H2,1H3,(H2,5,6,8)/i1D3
SMIL [2H]C([2H])([2H])N1CC(=O)N=C1N
IUPAC navn 2-amino-3-(trideuteriomethyl)-4H-imidazol-5-one
Molekylvægt (g/mol) 116.14
Formel vægt 116.0777
Procent renhed >95