missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine monohydrate, 99%, Thermo Scientific Chemicals
284.68 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Creatine monohydrate is involved in rapid ATP production primarily in skeletal muscle tissue via the action of creatine kinase(s). It may be used as a supplement to study its uptake mechanism and metabolism of action. It is used in the treatment of neuromuscular diseases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water (17 g/L) at 20°C. Insoluble in ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Soluble in water (17 g/L) at 20°C. Insoluble in ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications
Melting Point | ∼292°C (decomposition) |
pH | 6.9 |
Odor | Weak |
Quantity | 100 g |
Beilstein | 907175 |
Merck Index | 14,2568 |
Solubility Information | Soluble in water (17g/L) at 20°C. Insoluble in ether. |
Formula Weight | 149.15 (131.13 Anhydrous) |
Percent Purity | 99% |
Chemical Name or Material | Creatine monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'