Få mere at vide
Tocris Bioscience™ CRBN PROTAC™ 14a
Beskrivelse
CRBN PROTAC® 14a is a cereblon Degrader (PROTAC™). Comprises a ligand for Von Hippel Lindau (VHL) protein joined by a linker to a cereblon targeting ligand. Preferentially degrades cereblon (DC50 = 200 nM in HEK293 cells). Induces rapid and profound cereblon degradation (98% decrease after 8 h at 1 μM in HEK293 cells).
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N1-(2-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)ethyl)-N20-(-1-((2 S,4 R)-4-hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)-3,9,12,18-tetraoxaicosanediamide |
| CAS | 2358775-70-7 |
| Mængde | 5 mg |
| Mål | Active Degraders |
| Molekylær formel | C53H72N8O13S |
| Renhed | 0.98 |