Få mere at vide
Tocris Bioscience™ CRBN-6-5-5-VHL
Beskrivelse
Potent and selective cereblon Degrader (PROTAC™) (DC50 = 1.5 nM). Induces complete degradation of cereblon in MM1S cells. Comprises a cereblon E3 ligase ligand joined by a linker to a ligand for the E3 ligase VHL. Cell-permeable.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2 S,4 R)-1-((2 S)-2-(5-((5-((6-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)hexyl)oxy)pentyl)oxy)pentanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide |
| CAS | 2362575-45-7 |
| Mængde | 5 mg |
| Opløselighed | Soluble to 100mM in DMSO |
| Mål | Active Degraders |
| Molekylær formel | C51H69N7O10S |
| Formel vægt | 972.21 |
| Fysisk form | Solid |
| Farve | Yellow |
| Produkttype | Degrader |
| Vis mere |