missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRABP2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99740B
Additional Details : Weight : 0.00970kg
Description
CRABP2 Polyclonal specifically detects CRABP2 in Human samples. It is validated for Western Blot, ELISA.Specifications
CRABP2 | |
Polyclonal | |
Western Blot, ELISA | |
cellular retinoic acid binding protein 2, Cellular retinoic acid-binding protein II, CRABP-IIcellular retinoic acid-binding protein 2, RBP6 | |
Produced in rabbits immunized with purified, recombinant CRABP2 (NP_001869.1; Pro2-Glu 138). | |
0.1 mL | |
Core ESC Like Genes, Neuroscience, Stem Cell Markers | |
1382 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Biotin | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |