Få mere at vide
Tocris Bioscience™ CPI 613
PDH and KGDH inhibitor
Brand: Tocris Bioscience™ 5348/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
6,8-Bis(benzylthio)octanoic acid is a pyruvate dehydrogenase (PDH) and α-ketoglutarate dehydrogenase (KGDH) inhibitor. Disrupts tumor cell mitochondrial metabolism, and increases mitochondrial reactive oxygen species (ROS) production in H460 lung carcinoma cells. Has no effect on KGDH activity in normal bronchial epithelial cells. Induces cell death of multiple tumor cell lines selectively over normal cells in vitro. Inhibits tumor growth of H460 and BxPC-3 tumor cell xenografts in mice.Tekniske data
| 6,8-Bis[(phenylmethyl)thio]octanoic acid | |
| 95809-78-2 | |
| 50 mg |