Få mere at vide
CPI-203, MedChemExpress
Artikelnummer. 16483496
Encompass_Preferred
Skift visning
Mængde:
2 mg
5 mg
10 mg
25 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16483496 25 mg
16466868 10 mg
16459407 5 mg
16430266 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16483496 Leverandør Medchem Express Leverandørnr. HY15846
MedChemExpress CPI-203 is a novel potent, selective and cell permeable inhibitor of BET bromodomain, with an IC50 value of appr 37 nM (BRD4 α-screen assay).
CPI-203 is a novel potent, selective and cell permeable inhibitor of BET bromodomain, with an IC50 value of appr 37 nM (BRD4 α-screen assay).
Tekniske data
Kemisk navn eller materiale CPI-203
Fysisk form Solid
CAS 1446144-04-2
Farve Yellow
Mængde 25 mg
Renhedsgrad noter Research
Molekylær formel C19H18ClN5OS
Til brug med (applikation) Cancer-Kinase/protease
Opløselighedsinformation DMSO : ≥ 47 mg/mL (117.53 mM)
SMIL ClC1=CC=C(C(C2=C(N3C4=NN=C3C)SC(C)=C2C)=N[C@H]4CC(N)=O)C=C1
Molekylvægt (g/mol) 399.9
Formel vægt 399.9
Procent renhed 98.07%