Learn More
R&D Systems™ CpG oligodeoxynucleotides Mouse
Product Code. 18123255 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Product Code. Quantity Unit Size
18123255 0.1 mg 0.1mg
1 options
Product Code. 18123255 Supplier R&D Systems™ Supplier No. NBP231140
  • ONLINE EXCLUSIVE PRICE
    2061.49 DKK
    1827.00 DKK / 0.1mg
    Spare 234.49 DKK
    11% Slukket
 Restricted Item
Forventet forsendelse: 30-11-2026
A Novus product, part of R&D Systems.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Line Novus
Quantity 0.1 mg
For Use With (Application) Activation of mouse TLR9
Type CpG Oligodeoxynucleotides