Learn More
CPC™ Standard-Volume Quick-Disconnect Inline Hose-Barb Inserts: With Straight-Through Flow
With shut-off valve
137.00 DKK
Specifications
Product Type | Hose Barb Valved In-line Coupling Insert |
---|---|
Includes | Shut-off valves |
Maximum Pressure | 120 psi |
Product Code | Brand | Product Line | Material | MaxOperatingTempMetric | For Use With | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Line | Material | MaxOperatingTempMetric | For Use With | Price | Quantity & Availability | ||
11719649
|
CPC™
06364-80 |
PLC Series | EPDM Seal | 71°C | 6.4mm |
137.00 DKK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||
Description
- Flow sizes to 0.37 in. (9.5mm)
- Easy push-button or twist-off disconnection
- Chemical and corrosion resistant quick-disconnect bodies and inserts compatible with polyurethane, polyethylene, nylon, PVC, silicone and other tubing materials
- In white acetal or almond polypropylene, which can be mixed or matched for a variety of coupling types
- Type 316 stainless-steel metal parts (internal springs, valves or thumb latches)
- Ideal for use in laboratory equipment, medical and surgical instrumentation, water transfer, air systems and other applications
- Audible click is heard when coupling has been fully engaged; push-button or twist-off disconnection
- Complete coupling set requires both a body and an insert (sold separately)
- Note: Body and insert flow sizes must match, but tubing I.D. sizes can be different.
Body and insert flow sizes must match, but tubing I.D. sizes can be different.
Applications:
For use in laboratory equipment, medical and surgical instrumentation, water transfer, air systems and other applications
Specifications
Hose Barb Valved In-line Coupling Insert | |
Shut-off valves | |
120 psi |