Få mere at vide
Cole-Parmer™ CPC™ Sanitetsstik, PE-sleeve, Single-Cycle
Diameter (engelsk) Indre:
1/2 tomme
3/8 tommer
Pakningsstørrelse:
Hver
Beskrivelse
- Disposable single cycle sleeve.
- Save time and money on cleaning.
- Eliminates dead legs where bacteria can grow.
- Connects to most bio-bag connections.
- Steam-through connections will transform your steam-in place (SIP) process by making the connections cleaner, faster and easier.
- Innovative product design allows for a one-step process for sterilization and media transfer while eliminating dead spots, reducing the number of connections and eliminating the need for a laminar flow hood.
- The terminations on the fittings are set to industry standards for bio-reactors and popular sizes of flexible tubing, speeding the adaptation to existing systems and processes.
- Tear-away sleeve protects the fitting from actuation prior to complete sterilization.
- The polysulfone body and silicone membrane materials meet requirements of USP Class VI, and physicochemical tests.
Tekniske data
Tekniske data
| Oversigt | Sanitary x Barb |
| Diameter (engelsk) Indre | 3/8 tommer |
| Materiale | Polysulfone, PE Sleeve |
| Slangestørrelse | 3/4 in. |
| Slange Barb | 3/4 in. |
| Udløbsport | 3/4 in. |
| Steriliseringsmetode | Gamma or steam-in-place (SIP) |
| Produkttype | Sanitary Connector, Single-Cycle |