Learn More
Cole-Parmer™ CPC™ Polypropylene Straight-through Bulkhead Panel Mount Hose Barb
Quick Disconnect couplings feature a thumb latch for easy assembly
Brand:  Cole-Parmer™ 31303-28
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15487063
  • 167.00 DKK / Each
Estimated Shipment
04-12-2020
Add to basket
Description
Description
  • High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately)
  • Plug insert into coupling body to connect
  • Fitting halves lock into place immediately
  • To disconnect fitting halves, simply press the button on the coupling body
  • Valved models have a spring-operated 316 SS valve that seals the flow path when fitting halves are disengaged
  • All fitting bodies have 316 SS latch springs and EPDM seals
Specifications
Specifications
60 psi
Polypropylene
32°F
3/4 in.
3/8 in. Quick-Disconnect
Not Autoclavable
Straight-through Bulkhead Panel Mount Hose Barb
0°C
3/4 in.
Ethylene Oxide, Gamma