Learn More
Cole-Parmer™ CPC™ Panel-mount John Guest Valved Fitting
Quick-disconnect fitting with simple push-to-connect interface for your rigid tubing
Brand: Cole-Parmer™ 31304-50
Additional Details : Weight : 0.01000kg
Description
- Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
- Plug insert into body to connect fitting.
- Fitting halves lock into place immediately.
- To disconnect fitting halves, simply press the thumb latch on the coupling body.
Specifications
Fitting | |
180°F | |
Quick-Disconnect x Push-to-Connect | |
Autoclave or Ethylene Oxide (ETO) | |
Acetal |
White | |
82°C | |
1/4 x 1/4 in. | |
4.95 cm | |
120 psi |