Få mere at vide
CP 122721 hydrochloride, Tocris Bioscience™
High affinity NK1 receptor antagonist
Brand: Tocris Bioscience 3649/25
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CP 122721 hydrochloride is a high affinity NK1 antagonist (pIC50 = 9.8). Suppresses recruitment of immunocytes in respiratory syncytial virus (RSV)-infected rats after capsaicin stimulation. Also blocks substance P-induced excitation of locus ceruleus cells in guinea pig brain slices (IC50 = 7 nM). Exhibits anxiolytic and antidepressant-like effects. Active in vivo.Tekniske data
| 145877-52-7 | |
| (2S,3S)-N-[[2-Methoxy-5-(trifluoromethoxy)phenyl]methyl]-2-phenyl-3-piperidinamine dihydrochloride | |
| 25 mg |