Få mere at vide
CP 122721 hydrochloride, Tocris Bioscience™
GREENER_CHOICE
High affinity NK1 receptor antagonist
Brand:  Tocris Bioscience 3649/5
Encompass
Produktkode 15997890
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
CP 122721 hydrochloride is a high affinity NK1 antagonist (pIC50 = 9.8). Suppresses recruitment of immunocytes in respiratory syncytial virus (RSV)-infected rats after capsaicin stimulation. Also blocks substance P-induced excitation of locus ceruleus cells in guinea pig brain slices (IC50 = 7 nM). Exhibits anxiolytic and antidepressant-like effects. Active in vivo.
Tekniske data
Tekniske data
145877-52-7
(2S,3S)-N-[[2-Methoxy-5-(trifluoromethoxy)phenyl]methyl]-2-phenyl-3-piperidinamine dihydrochloride
5 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner