missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX4NB Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-00354F
Additional Details : Weight : 0.00970kg
Description
COX4NB Polyclonal specifically detects COX4NB in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
COX4NB | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C16orf2, C16orf4, chromosome 16 open reading frame 2, COX4 neighbor, COX4AL, FAM158Bchromosome 16 open reading frame 4, neighbor of COX4, NOC4family with sequence similarity 158, member B, Protein FAM158B | |
Produced in rabbits immunized with purified, recombinant COX4NB (O43402; Met1-Cys210). | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
10328 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |