Få mere at vide
Tocris Bioscience™ Covidcil-19
Beskrivelse
Covidcil-19 binds to the revised attenuator hairpin structure of the SARS-CoV-2 frameshifting element (FSE) with high affinity (Kd = 11 nM). It stabilizes the hairpin's folded state and reduces frameshifting efficiency in cells. Covidcil-19 inhibits viral propagation and reduces viral infectivity by > 3.5 orders of magnitude.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-[[4-(Methylamino)-2-quinazolinyl]amino]benzoic acid |
| CAS | 1225177-95-6 |
| Mængde | 50 mg |
| Mål | Antiviral Compounds |
| Molekylær formel | C16H14N4O2 |
| Renhed | 0.98 |