Få mere at vide
Epredia™ Dækglas, rektangler
Artikelnummer. 12765098 Shop alle Epredia produkter
Tykkelse (metrisk):
#1 (0,13 til 0,16 mm)
#1,5 (0,16 til 0,19 mm)
Dimensioner (L x B):
36 x 24 mm
64 x 48 mm
Pakningsstørrelse:
Hver
Pakke med 1000
Artikelnummer. 12765098
Brand: Epredia™ A67761330
Epredia™TM coverslips are ideally suited as cover material for specimens in microscopic examinations in medicine, biology and research. Manufactured from D 263TM M borosilicate glass from Schott AG. Available in various sizes and thicknesses.
  • Highly transparent and achromatic – practically free of any color distortion
  • Good planarity
  • Refraction index adjusted for microscopes
  • High optical homogeneity – practically free of inclusions, streaks, bubbles etc.
  • Very good chemical stability
TRUSTED_SUSTAINABILITY
Tekniske data
Form Rectangle
Længde (metrisk) 64 mm
Bredde (metrisk) 48 mm
Tykkelse (metrisk) #1 (0,13 til 0,16 mm)
Dimensioner (L x B) 64 x 48 mm
Mængde 100/Pk.