Learn More
Epredia™ Cover Slips, Rectangles
Product Code. 11936309 Shop All Epredia Products
Change view
Click to view available options
Thickness (Metric):
#1 (0.13 to 0.16 mm)
#1.5 (0.16 to 0.19 mm)
Dimensions (L x W):
36 x 24 mm
64 x 48 mm
Unit Size:
Each
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) Dimensions (L x W) Unit Size
11936309 #1.5 (0.16 to 0.19 mm) 36 x 24 mm Pack of 1000
12765098 #1 (0.13 to 0.16 mm) 64 x 48 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11936309 Supplier Epredia™ Supplier No. BB02400360AC13MNZ0
  • ONLINE EXCLUSIVE PRICE
    377.00 DKK
    334.82 DKK / Pakke med 1000
    Spare 42.18 DKK
    11% Slukket
 Restricted Item
Forventet forsendelse: 15-02-2027
Epredia™TM coverslips are ideally suited as cover material for specimens in microscopic examinations in medicine, biology and research. Manufactured from D 263TM M borosilicate glass from Schott AG. Available in various sizes and thicknesses.
  • Highly transparent and achromatic – practically free of any color distortion
  • Good planarity
  • Refraction index adjusted for microscopes
  • High optical homogeneity – practically free of inclusions, streaks, bubbles etc.
  • Very good chemical stability
TRUSTED_SUSTAINABILITY
Specifications
Shape Rectangle
Length (Metric) 36 mm
Width (Metric) 24 mm
Thickness (Metric) #1.5 (0.16 to 0.19 mm)
Dimensions (L x W) 36 x 24 mm
Quantity 100/Pk.