Learn More
Epredia™ Cover Slips, Circles
Product Code. 10343435 Shop All Epredia Products
Change view
Click to view available options
Thickness (Metric):
#0 (0.085 to 0.13 mm)
#1.5 (0.16 to 0.19 mm)
Diameter (Metric):
10 mm
30 mm
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) Diameter (Metric) Unit Size
10343435 #1.5 (0.16 to 0.19 mm) 30 mm Pack of 1000
12893804 #0 (0.085 to 0.13 mm) 10 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10343435 Supplier Epredia™ Supplier No. CB00300RAC40MNZ0
  • ONLINE EXCLUSIVE PRICE
    947.00 DKK
    840.28 DKK / Pakke med 1000
    Spare 106.72 DKK
    11% Slukket
 Restricted Item
Forventet forsendelse: 22-12-2026
Epredia™TM coverslips are ideally suited as cover material for specimens in microscopic examinations in medicine, biology and research. Manufactured from D 263TM M borosilicate glass from Schott AG. Available in various sizes thicknesses.
  • Highly transparent and achromatic – practically free of any color distortion
    Good planarity
  • Refraction index adjusted for microscopes
  • High optical homogeneity – practically free of inclusions, streaks, bubbles etc.
  • Very good chemical stability
TRUSTED_SUSTAINABILITY
Specifications
Shape Circle
Material Borosilicate Glass
Length (Metric) 30 mm
Width (Metric) 30 mm
Thickness (Metric) #1.5 (0.16 to 0.19 mm)
Diameter (Metric) 30 mm