Få mere at vide
Coumaphos, SPEX CertiPrep™
Artikelnummer. 12909494 Shop alle SPEX Certiprep produkter
Skift visning
Mængde:
1 mL
Pakningsstørrelse:
1ml
Artikelnummer. Mængde unitSize
12909494 1 mL 1ml
1 options
Artikelnummer. 12909494 Leverandør SPEX Certiprep Leverandørnr. S980
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Indeholder
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
Coumaphos56-72-41000μg/mL
Certificeringer
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Overholdelse
GHS/OSHA Complaint
Tekniske data
Kemisk navn eller materiale Coumaphos
Emballage 2mL Amber Prescored Glass Ampule
CAS 56-72-4, 67-64-1
CAS min % 0.1, 99.9
Koncentration eller sammensætningsnoter Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molekylær formel C14H16ClO5PS
Til brug med (udstyr) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI nøgle BXNANOICGRISHX-UHFFFAOYSA-N
SMIL CCOP(=S)(OCC)OC1=CC2=C(C=C1)C(=C(C(=O)O2)Cl)C
IUPAC navn 3-chloro-7-diethoxyphosphinothioyloxy-4-methylchromen-2-one
Opløsningsmiddel eller Matrix Acetone
Molekylvægt (g/mol) 362.761
PubChem CID 2871
Mængde 1 mL
ChEBI CHEBI:3903
Vis mere Vis mindre
Product intended for laboratory use only