Learn More
Cotton Blanket
Product Code. 12454940
Change view
Click to view available options
Dimensions (L x W):
150 x 200 cm
Unit Size:
Each
Product Code. Dimensions (L x W) Unit Size
12454940 150 x 200 cm Each
1 options
Product Code. 12454940 Supplier Safety First Supplier No. Q2024
  • ONLINE EXCLUSIVE PRICE
    189.57 DKK
    127.55 DKK / Pakke med 1
    Spare 62.02 DKK
    32% Slukket
Forventet forsendelse: 29-01-2027
Specifications
Product Type Blanket
Color Red
Material Cotton
Dimensions (L x W) 150 x 200 cm
Disposable Reusable
For Use With (Application) General use: to cover patients, to elevate limbs, to be used as a pillow, etc.
Length (Metric) 150 cm
Width (Metric) 200 cm