Learn More
Isofield™ Cos Non-Sterile Lightweight Cleanroom Sock - GMP Annex 1 Compatible
The Isofield Cos single-use non-sterile lightweight cleanroom sock is GMP Annex 1 compatible, offering comfort and protection in cleanroom settings. Available in three sizes, the Cos’ hourglass heel design ensures comfort levels rivalling most reusable alternatives.
5376.91 DKK
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
30178833
![]() |
Isofield™
70610S-M |
Small/Medium |
5376.91 DKK
500 pairs |
Estimated Shipment: 19-08-2025
Log in to see stock. |
||||||
30178852
![]() |
Isofield™
70610L-XL |
Large/X-Large |
5376.91 DKK
500 pairs |
Estimated Shipment: 19-08-2025
Log in to see stock. |
||||||
Description
- Non-sterile lightweight
- Cleanroom socks
- Single-use
- 95% polyester, 5% spandex
- Mid-calf length
- Low linting
- White color
Specifications
Cleanroom Socks |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Isofield™ Cos Non-Sterile Lightweight Cleanroom Sock - GMP Annex 1 Compatible