missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Corylifol A, MedChemExpress
Beskrivelse
Corylifol A inhibits IL-6-induced STAT3 activation and phosphorylation, with an IC50 of 0.81 μM.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Corylifol A |
| CAS | 775351-88-7 |
| Farve | White |
| Renhedsgrad noter | Research |
| Molekylær formel | C25H26O4 |
| Mængde | 5 mg |
| Synonym | Corylifol-A Corylinin |
| Opløselighedsinformation | DMSO : 250 mg/mL (640.25 mM; Need ultrasonic) |
| SMIL | O=C1C(C2=CC=C(O)C(C/C=C(C)/CC/C=C(C)\C)=C2)=COC3=CC(O)=CC=C13 |
| Molekylvægt (g/mol) | 390.47 |
| Vis mere |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?