Få mere at vide
Cortisol-9,12,12-d3, TRC
Artikelnummer. 30403256
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30403256 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403256 Leverandør Toronto Research Chemicals Leverandørnr. TRCH7146245MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cortisol-9,12,12-d3
CAS 115699-92-8
Molekylær formel C21 D3 H27 O5
Mængde 5 mg
Synonym (11β)-11,17,21-Trihydroxy-pregn-4-ene-3,20-dione-d3, (11β)-11,17,21-Trihydroxypregn-4-ene-3,20-dione-9,12,12-d3, Cortisol-d3, 11β,17,21-Trihydroxypregn-4-ene-3,20-dione-d3, 17-Hydroxycorticosterone-d3, 17α-Hydroxycorticosterone-d3, NSC 10483-d3, Hydrocortisone D3 (9,12,12 D3), Cortisol D3 (9,12,12 D3)
InChI formel InChI=1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1/i10D2,18D
SMIL [2H]C1([2H])[C@H](O)[C@@]2([2H])[C@@H](CCC3=CC(=O)CC[C@]23C)[C@@H]4CC[C@](O)(C(=O)CO)[C@@]14C
IUPAC navn (8S,9S,10R,11S,13S,14S,17R)-9,12,12-trideuterio-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,11,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-3-one
Molekylvægt (g/mol) 365.48
Formel vægt 365.2282