Få mere at vide
Cortisol-9,12,12-d3, CDN
Artikelnummer. 30310832
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30310832 0.01 g
30309634 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310832 Leverandør CDN Leverandørnr. CDND80980.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cortisol-9,12,12-d3
CAS 115699-92-8
Molekylær formel C21 D3 H27 O5
Mængde 0.01 g
Synonym (11β)-11,17,21-Trihydroxy-pregn-4-ene-3,20-dione-d3, (11β)-11,17,21-Trihydroxypregn-4-ene-3,20-dione-9,12,12-d3, Cortisol-d3, 11β,17,21-Trihydroxypregn-4-ene-3,20-dione-d3, 17-Hydroxycorticosterone-d3, 17α-Hydroxycorticosterone-d3, NSC 10483-d3, Hydrocortisone D3 (9,12,12 D3), Cortisol D3 (9,12,12 D3)
InChI formel InChI=1 S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26 H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1/i10D2,18 D
SMIL [2 H]C1([2 H])[C@H](O)[C@@]2([2 H])[C@@H](CCC3=CC(=O)CC[C@]23 C)[C@@H]4 CC[C@](O)(C(=O)CO)[C@@]14 C
IUPAC navn (8 S,9 S,10 R,11 S,13 S,14 S,17 R)-9,12,12-trideuterio-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,11,14,15,16-octahydro-1 H-cyclopenta[a]phenanthren-3-one
Molekylvægt (g/mol) 365.48
Formel vægt 365.2282 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity