Få mere at vide
Cortisol 21-Sulfate Potassium Salt, TRC
Artikelnummer. 30136523
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136523 250 mg
30004054 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136523 Leverandør Toronto Research Chemicals Leverandørnr. TRCC696320250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 152957-32-9
Molekylær formel C21H29KO8S
Molekylvægt (g/mol) 480.61
Synonym Pregn-4-ene-3,20-dione, 11,17-dihydroxy-21-(sulfooxy)-, monopotassium salt, (11β)- (9CI), (11β)-11,17-Dihydroxy-21-(sulfooxy)pregn-4-ene-3,20-dione monopotassium salt, Potassium hydrocortisone 21-sulfate, Cortisol 21-sulfate potassium salt
IUPAC navn potassium;[2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] sulfate
SMIL [K+].C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)COS(=O)(=O)[O-])[C@@]4(C)C[C@H](O)[C@H]23
Tekniske data
Kemisk navn eller materiale Cortisol 21-Sulfate Potassium Salt
Mængde 250 mg
InChI formel InChI=1S/C21H30O8S.K/c1-19-7-5-13(22)9-12(19)3-4-14-15-6-8-21(25,17(24)11-29-30(26,27)28)20(15,2)10-16(23)18(14)19;/h9,14-16,18,23,25H,3-8,10-11H2,1-2H3,(H,26,27,28);/q;+1/p-1/t14-,15-,16-,18+,19-,20-,21-;/m0./s1
Formel vægt 480.122