Få mere at vide
Cortisol 21-Beta-D-Glucuronide Sodium Salt(>90%), TRC
Artikelnummer. 30011541
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011541 1 mg
30009596 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011541 Leverandør Toronto Research Chemicals Leverandørnr. TRCC6963151MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 213456-71-4
Molekylær formel C27 H37 O11 . Na
Molekylvægt (g/mol) 560.57
Synonym β-D-Glucopyranosiduronic acid, (11β)-11,17-dihydroxy-3,20-dioxopregn-4-en-21-yl, monosodium salt (9CI), Sodium cortisol 21-β-D-glucuronide
IUPAC navn sodium;(2S,3S,4S,5R,6R)-6-[2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]-3,4,5-trihydroxyoxane-2-carboxylate
SMIL [Na+].C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)[O-])[C@@]4(C)C[C@H](O)[C@H]23
Tekniske data
Kemisk navn eller materiale Cortisol 21-beta-D-Glucuronide sodium salt
Mængde 1 mg
InChI formel InChI=1S/C27H38O11.Na/c1-25-7-5-13(28)9-12(25)3-4-14-15-6-8-27(36,26(15,2)10-16(29)18(14)25)17(30)11-37-24-21(33)19(31)20(32)22(38-24)23(34)35;/h9,14-16,18-22,24,29,31-33,36H,3-8,10-11H2,1-2H3,(H,34,35);/q;+1/p-1/t14-,15-,16-,18+,19-,20-,21+,22-,24+,25-,26-,27-;/m0./s1
Formel vægt 560.2234