missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
MedChemExpress Corticotropin-releasing factor (human)
Specifications
Specifications
| Renhed | 98.44% |
| CAS | 86784-80-7 |
| Indhold og opbevaring | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| Til brug med (applikation) | Neuroscience-Neuromodulation |
| Molekylvægt (g/mol) | 4757.45 g/mol |
| Produkttype | Peptides |
| Mængde | 250 μg |
| Forsendelsestilstand | Room temperature |
| Opløselighed | H2O : 25 mg/mL (5.25 mM∣ultrasonic and adjust pH to 2 with HCl) |
| Synonym | Human CRF∣ Human corticotropin-releasing factor |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?