Få mere at vide
Coronene-d12, TRC
Artikelnummer. 17989229
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17989229 10 mg
17978119 5 mg
17908629 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 17989229 Leverandør Toronto Research Chemicals Leverandørnr. TRCC69550210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 16083-32-2
Molekylær formel C24 2H12
Molekylvægt (g/mol) 312.43
Synonym Coronene-d12 (8CI,9CI), Coronene D12
IUPAC navn 1,2,3,4,5,6,7,8,9,10,11,12-dodecadeuteriocoronene
SMIL [2H]c1c([2H])c2c([2H])c([2H])c3c([2H])c([2H])c4c([2H])c([2H])c5c([2H])c([2H])c6c([2H])c([2H])c1c7c6c5c4c3c27
Tekniske data
Kemisk navn eller materiale Coronene-d12
Mængde 10 mg
InChI formel InChI=1S/C24H12/c1-2-14-5-6-16-9-11-18-12-10-17-8-7-15-4-3-13(1)19-20(14)22(16)24(18)23(17)21(15)19/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D
Formel vægt 312.1692