Learn More
Corning™ PureCoat™ Carboxyl Surface Dishes
Negatively charged surface improves attachment. increases proliferation and improved differentiation for a broad range of cells that exhibit poor attachment properties in serum-free or serum-reduced conditions
Brand:  Corning™ 354784
Additional Details : Weight : 0.38000kg
 View more versions of this product
Product Code. 11503590
  • 309.00 DKK / Pack of 10
Estimated Shipment
07-06-2021
Add to basket
Description
Description
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Specifications
Specifications
Flat
58.1 cm2
Polystyrene
Cell Culture Dish
10/Pk.
Round
100 mm
Carboxyl
10/Cs.