Learn More
Corning™ X-GAL, Powder
Produced under highest industry standards
Brand:  Corning™ 46-101-RF
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15383721
  • 2075.00 DKK / 1g
Estimated Shipment
28-05-2021
Add to basket
Description
Description
X-Gal (5-bromo-4-chloro-3-indolyl-b-D-galactopyranoside)
  • Substrate of b-galactosidase
  • Hydrolyzed by β-galactosidase to produce indoxyl in bacterial medium
Specifications
Specifications
5-bromo-4-chloro-3-indolyl-b-D-galactopyranoside
Research use only
1000 mg