missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Medium 199 (Mod.)

Often used in vaccine production, virology, and for the culture of various non-transformed cells. It was originally formulated as the result of efforts to create a fully defined, animal origin-free medium.

Brand:  Corning™ 90-050-PB

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 15373781

  • 615.00 DKK / 10 L

Estimated Shipment
04-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Product manufactured under industry-leading standards for low endotoxin content and superior results. Medium 199 is often used in vaccine production, virology, and for the culture of various non-transformed cells.  It was originally formulated as the result of efforts to create a fully defined, animal origin-free medium. Long-term use of this medium requires serum supplementation. This formulation contains Earle's salts and L-glutamine, and it does not contain sodium bicarbonate.
Specifications

Specifications

10000 mL
Sodium Bicarbonate, L-Glutamine, Phenol Red
2° to 8°C
With Earle's Salts
2° to 8°C
Videos
SDS
Documents