Learn More
Proteintech CoraLite™488-Annexin V and PI Apoptosis Kit
CoraLite™488-Annexin V and PI Apoptosis Detection Kit provides a quick and easy method to detect apoptosis by marking early apoptotic cells (green) and late apoptotic cells or necrotic cells (red).
Brand:  Proteintech PF00005-50T
Additional Details : Weight : 0.01030kg
Product Code. 17378252
  • 782.47 DKK / 50 tests
Estimated Shipment: 18-06-2024
to see stock.
Description
Description
  • Results can be read out using flow cytometry or other fluorescence testing equipment.
  • Early apoptic cells are identified by annexin V (labeled with CoraLite(R) 488 dye) binding to phosphatidylserine (PS) flipped to the extracellular membrane surface, an early event in apoptosis.
  • Compared with FITC, CoraLite(R) 488 dye has higher fluorescence brightness, and is not affected by pH in the environment, and has good light stability.