Learn More
Copper(II) Sulfate Pentahydrate, Supelco™
Product Code. 15847130
Change view
Click to view available options
Quantity:
5 kg
50 kg
Unit Size:
50kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15847130 50 kg 50kg
15837130 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15847130 Supplier Supelco Supplier No. 1027809050
 Restricted Item
Forventet forsendelse: 19-11-2026
Very fine crystals EMPLURA™
very fine crystals EMPLURA™
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7758-99-8
Specifications
Melting Point 147°C
Quantity 50 kg
MDL Number MFCD00149681
Synonym Copper(II) sulfate pentahydrate, Copper(II) sulfate pentahydrate
Molecular Weight (g/mol) 249.68
Grade Research
Density 2.284 g/cm3 at 20°C
Chemical Name or Material Copper(II) Sulfate Pentahydrate