missing translation for 'onlineSavingsMsg'
Learn More

Copper(Ii) Sulfate Pentahydrate, ACS Reagent, Honeywell Fluka™

Catalog Number 15641890
Click to view available options
Quantity:
100 g
250 g
500 g
2.5 kg
This item is not returnable. View return policy
7758-99-8
CuH10O9S
249.68
MFCD00149681
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
24463
CHEBI:31440
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
7758-99-8
CuH10O9S
249.68
MFCD00149681
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
24463
CHEBI:31440
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 250 g
Linear Formula CuSO4 · 5H2O
UN Number 3077
IUPAC Name copper;sulfate;pentahydrate
Formula Weight 249.69g/mol
Percent Purity ≥98%
Grade ACS Reagent
Vapor Pressure 7.3mmHg (25°C)
Chemical Name or Material Copper(Ii) Sulfate Pentahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'