Få mere at vide
Copper(II) Oxide, TRC
Artikelnummer. 30135099
Skift visning
Mængde:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135099 50 g
30137857 25 g
30011741 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135099 Leverandør Toronto Research Chemicals Leverandørnr. TRCC68535050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1317-38-0
Molekylær formel CuO
Molekylvægt (g/mol) 79.55
Synonym BYK-LP x 20704, Banacobru OL, Black copper oxide, C.I. 77403, C.I. Pigment Black 15, Coopers Permatrace Copper, Copacaps, Copinox Lamb, Copper Brown, Copper monooxide, Copper monoxide, Copper monoxide (CuO), Copper oxide, Copper oxide (Cu4O4), Copper(2+) oxide, Copper(II) oxide, Copper(II) oxide (CuO), Copporal, Cupric oxide, ET, N 120, N 120 (oxide), N 520, NSC 83537, NanoActive CuO, NanoArc U 1102DBE, Nanotek CuO, Nanotek CuO-C, YC
IUPAC navn oxocopper
SMIL O=[Cu]
Tekniske data
Kemisk navn eller materiale Copper(II) oxide
Mængde 50 g
InChI formel InChI=1S/Cu.O
Formel vægt 78.9245