missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxide, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407440
Click to view available options
Quantity:
25 g
100 g
500 g
1317-38-0
CuO
79.55
MFCD00010979
KKCXRELNMOYFLS-UHFFFAOYSA-N
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
14829
oxocopper
[O--].[Cu++]
1317-38-0
CuO
79.55
MFCD00010979
KKCXRELNMOYFLS-UHFFFAOYSA-N
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
14829
oxocopper
[O--].[Cu++]

It is used as an adsorbent and absorbent, pigment, antifouling agent, wood preservative, optical glass polishing agent, welding flux for bronze materials, agricultural chemical and abrasive. It is used in the preparation of intermediates, oxidizing/reducing agents, surface treating agents and process regulators. It finds use in the manufacture of rayons, phosphors, and dry battery cells. It serves as a catalyst in certain chemical reactions of industrial importance. It is also employed in chemical sensors and superconducting materials. Hazardous materials like cyanides, dioxins and halogenated hydrocarbons can be disposed through oxidation with copper oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1326°C
Color Black
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.995% (Metal basis)
Solubility Information Practically insoluble in water,and alcohol; soluble in dilute acids,alkali cyanides,and ammonium carbonate solution.
Formula Weight 79.55
Odor Odorless
Refractive Index 2.63
Appearance Black powder
Density 6.3
Chemical Name or Material Copper(II) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'