missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) nitrate hemi(pentahydrate), ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog Number p-3256989
Click to view available options
Quantity:
50 g
500 g
2 kg
10 kg
This item is not returnable. View return policy
19004-19-4
CuN2O6·2.5H2O
MFCD00149671
dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate
This item is not returnable. View return policy
19004-19-4
CuN2O6·2.5H2O
MFCD00149671
dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate

For Use As a Catalyst, Nitrating Agent For Aromatic organosilicon Compounds, Nickel-Plating Baths

Catalyst, nitrating agent for aromatic organosilicon compounds, nickel-plating baths, coloring copper black and producing a burnished effect on ironCopper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 170°C (decomposition)
Melting Point ∽115°C
Physical Form Crystalline Lump
Quantity 50 g
Assay Percent Range 98.0-102.0%
Solubility Information Soluble in water and alcoholSoluble in water,ammonia and ethanol.
Formula Weight 232.59 (187.55 Anhydrous)
Grade ACS
Sensitivity Hygroscopic
Density 2.32 g/cm3
Chemical Name or Material Copper(II) nitrate hemi(pentahydrate), trihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'