missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) formate tetrahydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
It is employed as an antibacterial agent in the treatment of cellulose.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as an antibacterial agent in the treatment of cellulose.
Solubility
Soluble in water (125 g/L at 20°C). Very slightly soluble in alcohol. Practically insoluble in most organic solvents.
Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is employed as an antibacterial agent in the treatment of cellulose.
Solubility
Soluble in water (125 g/L at 20°C). Very slightly soluble in alcohol. Practically insoluble in most organic solvents.
Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Quantity | 50 g |
UN Number | UN3077 |
Merck Index | 14,2639 |
Solubility Information | Soluble in water (125g/L at 20°C). Very slightly soluble in alcohol. Practically insoluble in most organic solvents. |
Formula Weight | 225.65 (153.59 Anhydrous) |
Percent Purity | 98% |
Chemical Name or Material | Copper(II) formate tetrahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'