Learn More
Copper(II) carbonate basic, Puriss., 54-57% Cu basis, green, Honeywell Fluka™
Brand:  Honeywell Fluka 12830-1KG
Additional Details : CAS Number : 12069-69-1 Weight : 1.00000kg
Quantity 1kg
 View more versions of this product
Product Code. 15641200
  • 1212.75 DKK / 1kg
Estimated Shipment date
from Supplier 27-07-2018
Add to basket
Specifications
Chemical Identifiers
12069-69-1
Copper(Ii) Carbonate Basic
puriss.
Specifications
Copper(Ii) Carbonate Basic
puriss.
221.12g/mol
NONH for all modes of transport
Green
12069-69-1
CuCO3 · Cu(OH)2
MFCD00010976
54 to 57% Cu basis
1kg
Videos